MilliporeSigma
A method for synthesis of homoallylic bromide.
The Journal of organic chemistry (2013-05-22)
Wenke Qi, Peipei Wang, Liyuan Fan, Songlin Zhang
ABSTRACT
Cyclopropyl Grignard reagents react with carbonyl compounds in the presence of diethyl phosphite to give homoallylic bromides. The reaction is effectively carried out under mild conditions in a one-pot fashion with moderate to good yields.
Sigma-Aldrich
Diethyl phosphite, technical grade, 94%