MilliporeSigma
  • An assembly chaperone collaborates with the SMN complex to generate spliceosomal SnRNPs.
An assembly chaperone collaborates with the SMN complex to generate spliceosomal SnRNPs.
Cell (2008-11-06)
Ashwin Chari, Monika M Golas, Michael Klingenhäger, Nils Neuenkirchen, Bjoern Sander, Clemens Englbrecht, Albert Sickmann, Holger Stark, Utz Fischer
ABSTRACT
Spliceosomal small nuclear ribonucleoproteins (snRNPs) are essential components of the nuclear pre-mRNA processing machinery. A hallmark of these particles is a ring-shaped core domain generated by the binding of Sm proteins onto snRNA. PRMT5 and SMN complexes mediate the formation of the core domain in vivo. Here, we have elucidated the mechanism of this reaction by both biochemical and structural studies. We show that pICln, a component of the PRMT5 complex, induces the formation of an otherwise unstable higher-order Sm protein unit. In this state, the Sm proteins are kinetically trapped, preventing their association with snRNA. The SMN complex subsequently binds to these Sm protein units, dissociates pICln, and catalyzes ring closure on snRNA. Our data identify pICln as an assembly chaperone and the SMN complex as a catalyst of spliceosomal snRNP formation. The mode of action of this combined chaperone/catalyst system is reminiscent of the mechanism employed by DNA clamp loaders.