MilliporeSigma
  • The structural and functional heterogeneity of glutamic acid decarboxylase: a review.
The structural and functional heterogeneity of glutamic acid decarboxylase: a review.
Neurochemical research (1991-03-01)
M G Erlander, A J Tobin
ABSTRACT
Studies of the GABA-synthetic enzyme glutamate decarboxylase (glutamic acid decarboxylase; GAD; E.C.4.1.1.15) began in 1951 with the work of Roberts and his colleagues. Since then, many investigators have demonstrated the structural and functional heterogeneity of brain GAD. At least part of this heterogeneity derives from the existence of two GAD genes.