MilliporeSigma
  • Disulfide bond formation and activation of Escherichia coli β-galactosidase under oxidizing conditions.
Disulfide bond formation and activation of Escherichia coli β-galactosidase under oxidizing conditions.
Applied and environmental microbiology (2012-01-31)
Joaquin Seras-Franzoso, Roman Affentranger, Mario Ferrer-Navarro, Xavier Daura, Antonio Villaverde, Elena García-Fruitós
ABSTRACT
Escherichia coli β-galactosidase is probably the most widely used reporter enzyme in molecular biology, cell biology, and biotechnology because of the easy detection of its activity. Its large size and tetrameric structure make this bacterial protein an interesting model for crystallographic studies and atomic mapping. In the present study, we investigate a version of Escherichia coli β-galactosidase produced under oxidizing conditions, in the cytoplasm of an Origami strain. Our data prove the activation of this microbial enzyme under oxidizing conditions and clearly show the occurrence of a disulfide bond in the β-galactosidase structure. Additionally, the formation of this disulfide bond is supported by the analysis of a homology model of the protein that indicates that two cysteines located in the vicinity of the catalytic center are sufficiently close for disulfide bond formation.
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein
Pricing and availability is not currently available.