MilliporeSigma
  • Clustered arrangement of winged helix genes fkh-6 and MFH-1: possible implications for mesoderm development.
Clustered arrangement of winged helix genes fkh-6 and MFH-1: possible implications for mesoderm development.
Development (Cambridge, England) (1996-06-01)
K H Kaestner, S C Bleckmann, A P Monaghan, J Schlöndorff, A Mincheva, P Lichter, G Schütz
ABSTRACT
The 'winged helix' or 'forkhead' transcription factor gene family is defined by a common 100 amino acid DNA binding domain which is a variant of the helix-turn-helix motif. Here we describe the structure and expression of the mouse fkh-6 and MFH-1 genes. Both genes are expressed in embryonic mesoderm from the headfold stage onward. Transcripts for both genes are localised mainly to mesenchymal tissues, fkh-6 mRNA is enriched in the mesenchyme of the gut, lung, tongue and head, whereas MFH-1 is expressed in somitic mesoderm, in the endocardium and blood vessels as well as the condensing mesenchyme of the bones and kidney and in head mesenchyme. Both genes are located within a 10 kb region (in mouse chromosome 8 at 5.26 +/- 2.56 cM telomeric to Actsk1. The close physical linkage of these two winged helix genes is conserved in man, where the two genes map to chromosome 16q22-24. This tandem arrangement suggests the common use of regulatory mechanisms. The fkh-6/MFH-1 locus maps close to the mouse mutation amputated, which is characterised by abnormal development of somitic and facial mesoderm. Based on the expression patterns we suggest that a mutation in MFH-1, not fkh-6 is the possible cause for the amputated phenotype.