MilliporeSigma
  • Studies on pyrazine derivatives. XLVII. Synthesis and antibacterial activity of novel pyrazine derivatives with amidoxime moiety.
Studies on pyrazine derivatives. XLVII. Synthesis and antibacterial activity of novel pyrazine derivatives with amidoxime moiety.
Acta poloniae pharmaceutica (2007-05-23)
Katarzyna Gobis, Henryk Foks, Anna Kedzia, Maria Wierzchowska, Ewa Kwapisz, Zofia Zwolska, Ewa Augustynowicz-Kopeć
ABSTRACT
The new pyrazine derivatives exhibiting an antibacterial activity have been synthesized. Initial amidoxime 1 was obtained in the reaction of pyrazinecarbonitrile with hydroxylamine. Upon treatment of amidoxime with methyl iodide O-methyl derivative 2 was formed. Both amidoximes were transformed into imidoyl chlorides 3, 4. Then the chloride atom in those derivatives was substituted with various secondary amines giving appropriate oximes 5-18 and O-methyl-oximes 19 and 20. The obtained compounds were tested in vitro for their tuberculostatic activity. The inhibiting concentration (MIC) values were within 25-100 microg/mL. Their activity towards 25 strains of anaerobic and 25 strains of aerobic bacteria was also studied. Three compounds exhibited activity against both types of bacteria.
Sigma-Aldrich
Pyrazinecarbonitrile, 99%