MilliporeSigma
Inhibition of noroviruses by piperazine derivatives.
Bioorganic & medicinal chemistry letters (2011-11-29)
Dengfeng Dou, Guijia He, Sivakoteswara Rao Mandadapu, Sridhar Aravapalli, Yunjeong Kim, Kyeong-Ok Chang, William C Groutas
ABSTRACT
There is currently an unmet need for the development of small-molecule therapeutics for norovirus infection. The piperazine scaffold, a privileged structure embodied in many pharmacological agents, was used to synthesize an array of structurally-diverse derivatives which were screened for anti-norovius activity in a cell-based replicon system. The studies described herein demonstrate for the first time that functionalized piperazine derivatives possess anti-norovirus activity. Furthermore, these studies have led to the identification of two promising compounds (6a and 9l) that can be used as a launching pad for the optimization of potency, cytotoxicity, and drug-like characteristics.
Sigma-Aldrich
1-Boc-piperazine, 97%