MilliporeSigma
Expression of huCdc7 in colorectal cancer.
World journal of gastroenterology (2013-05-30)
Hai-Jun Chen, Zhen Zhu, Xue-Lin Wang, Quan-Lin Feng, Qing Wu, Zheng-Ping Xu, Jiang Wu, Xiao-Feng Yu, Hong-Liang Qian, Qi Lu
ABSTRACT
To detect the expression of huCdc7 in colorectal cancer. The mRNA and protein expression of huCdc7 in 39 colorectal cancer tissue specimens and matched tumor-adjacent normal colorectal tissue specimens was detected by reverse transcription-polymerase chain reaction and immunohistochemistry, respectively. The relative expression level of huCdc7 mRNA in colorectal cancer was significantly higher than that in tumor-adjacent normal colorectal tissues (0.03675 ± 1.00 vs 0.01199 ± 0.44, P < 0.05). huCdc7-positive cells displayed brown granules in the nucleus. Tumor tissues contained many huCdc7-positive cells, whereas normal colorectal tissues contained very few positive cells. huCdc7 may play an important role in the development and progression of colorectal cancer.