MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • 1,5-Benzoxazepines vs 1,5-benzodiazepines. one-pot microwave-assisted synthesis and evaluation for antioxidant activity and lipid peroxidation inhibition.
1,5-Benzoxazepines vs 1,5-benzodiazepines. one-pot microwave-assisted synthesis and evaluation for antioxidant activity and lipid peroxidation inhibition.
Journal of medicinal chemistry (2010-11-06)
Constantinos G Neochoritis, Constantinos A Tsoleridis, Julia Stephanidou-Stephanatou, Christos A Kontogiorgis, Dimitra J Hadjipavlou-Litina
ABSTRACT
Amino-1,5-benzoxazepines 2 and 5 and hydroxyl-1,5-benzodiazepines 3 and 6 have been synthesized in one-pot solvent-free conditions from 2,3-diaminophenol and ketones through microwave assisted acid catalysis, the benzoxazepine/benzodiazepine ratio depending on the R(1) and R(3) aryl substituents. The otherwise inaccessible and unknown 2,2-dimethyl-4-aryl-1,5-benzodiazepines 8 were also prepared in an analogous manner. The reaction mechanism was investigated by means of DFT calculations. Structural assignments of the new compounds as well as complete assignment of (1)H and (13)C NMR signals have been unambiguously achieved on the basis of the analysis of their (1)H and (13)C NMR (1D and 2D), IR, MS, and elemental analysis data, whereas the presence of an amino group in 5 and of a hydroxyl in 6 was confirmed by derivatization. Compounds 2, 3, 5f, 6a, 6c, 6d, 6f, 6h, 8c, and 12 were evaluated as antioxidants and lipid peroxidation inhibitors in vitro. Compound 6f was also evaluated as anti-inflammatory agent in vivo. Compounds 2 and 6f were found to be the most potent as inhibitors of lipoxygenase and of lipid peroxidation, respectively.
Sigma-Aldrich
2,3-Diaminophenol, 97%