MilliporeSigma
  • Extracellular polymeric substances and bound water drastically affect bioleached sludge dewaterability at low temperature.
Extracellular polymeric substances and bound water drastically affect bioleached sludge dewaterability at low temperature.
Environmental technology (2014-08-26)
Yongwei Song, Guanyu Zheng, Minbo Huo, Bowen Zhao, Lixiang Zhou
ABSTRACT
It is well documented that bioleaching with the co-inoculation ofAcidithiobacillus ferrooxidans and Acidithiobacillus thiooxidans can drastically enhance sludge dewaterability under ambient temperature condition, but little information on low temperature effect on bioleached sludge dewaterability is available. In this study, the optimum sludge retention time of bioleaching treatment and the mechanisms responsible for the dewaterability enhancement ofbioleached sludge were studied in two bioleaching systems conducted at a low temperature of 10 degrees C and a normal temperature of 28 degrees C, respectively. Results showed that Acidithiobacillus bacteria involved in the bioleaching were still active at 10 degrees C, and sludge capillary suction time (CST) decreased in the first 3 days of bioleaching. Nevertheless, the degree of sludge dewaterability enhancement by bioleaching was much lower at 10 degrees C than that at 28 degrees C. The only slight decreases in the sludge pH value and relative high contents of extracellular polymeric substances (EPS) and bound water at a low temperature were responsible for the relatively lower dewaterability enhancement of bioleached sludge compared to that at the normal temperature. Moreover, statistical results showed that sludge CST had significant positive correlations with the sludge pH value, and EPS and bound water contents in the optimum bioleaching process (from day 0 to day 3). Therefore, reducing medium pH, and/or removing EPS from bioleaching bacteria and bound water ofbioleached sludge through forced physical-chemical approaches prior to mechanical dewatering might be a good method to greatly improve sludge dewatering at a low temperature.
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, foil, 2m coil, thickness 0.125mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.025mm, hard, 99.5%
Iron, foil, 2m coil, thickness 0.15mm, hard, 99.5%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 2m coil, thickness 0.38mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.38mm, hard, 99.5%
Iron, foil, 2m coil, thickness 0.038mm, hard, 99.5%
Iron, wire reel, 10m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.005mm, 99.85%
Iron, foil, not light tested, 100x100mm, thickness 0.0025mm, 99.85%
Iron, foil, 5m coil, thickness 0.5mm, coil width 49mm, armco« soft ingot 99.8+%
Iron, foil, light tested, 100x100mm, thickness 0.025mm, hard, 99.5%
Iron, rod, 1000mm, diameter 2.0mm, as drawn, 99.99+%
Iron, foil, not light tested, 25x25mm, thickness 0.002mm, 99.85%
Iron, foil, not light tested, 100x100mm, thickness 0.007mm, 99.85%
Iron, foil, not light tested, 300x300mm, thickness 0.05mm, hard, 99.5%
Iron, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, light tested, 100x100mm, thickness 0.038mm, hard, 99.5%
Iron, foil, not light tested, 25x25mm, thickness 0.003mm, 99.85%
Iron, microfoil, disks, 10mm, thinness 0.05μm, specific density 40.44μg/cm2, permanent mylar 3.5μm support, 99.99+%
Iron, foil, not light tested, 150x150mm, thickness 0.009mm, 99.85%
Iron, rod, 100mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Iron, rod, 1000mm, diameter 19.0mm, as drawn, 98+%
Iron, rod, 100mm, diameter 25mm, as drawn, 98+%