MilliporeSigma
The toxic effects of silver nanoparticles on blood mononuclear cells.
The international journal of occupational and environmental medicine (2014-07-17)
A Barkhordari, S Barzegar, H Hekmatimoghaddam, A Jebali, S Rahimi Moghadam, N Khanjani
ABSTRACT
Nanoparticles have become one of the leading technologies over the past two years. The extensive use of nanoparticles has raised great concern about their occupational fate and biological effects. With an increase in the production and use of nanomaterial, it is more likely to get exposed to them occupationally and environmentally. To assess the toxicity of silver nanoparticles on human mononuclear cells. In this in vitro experimental study, suspensions of blood mononuclear cells from 10 young healthy men were incubated with 10-nm silver nanoparticles in different concentrations (range: 1-500 μg/mL) for 6 and 24 hours by MTT assay. Positive and negative controls were used for comparison. After 6 hours of exposure, 10.9% to 48.4% of the cells died. After 24 hours of exposure, the rate ranged from 56.8% to 86.3%. Regardless of the exposure time, the maximum cytotoxicity was observed at the concentration of 500 μg/mL of silver nanoparticles. By increasing the exposure time to 24 hours, the cytotoxicity of nanoparticles substantially increased at all concentrations. Cell death was significantly higher when compared to the controls (p<0.01). Silver nanoparticles possess both time- and dose-dependent cytotoxicity and can thus be considered as very toxic for mononuclear cells.
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 200  nm × 10 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, conductive paste
Silver, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95+%
Silver, foil, 15mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95+%
Silver, foil, 4mm disks, thickness 0.007mm, 99.97%
Silver, foil, 15mm disks, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.015mm, 99.97%
Silver, foil, 1m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 1m coil, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.01mm, 99.95+%
Silver, foil, 1m coil, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 2m coil, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95+%