MilliporeSigma
  • Progressive disorganization of paranodal junctions and compact myelin due to loss of DCC expression by oligodendrocytes.
Progressive disorganization of paranodal junctions and compact myelin due to loss of DCC expression by oligodendrocytes.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-07-18)
Sarah-Jane Bull, Jenea M Bin, Eric Beaumont, Alexandre Boutet, Paul Krimpenfort, Abbas F Sadikot, Timothy E Kennedy
ABSTRACT
Paranodal axoglial junctions are critical for maintaining the segregation of axonal domains along myelinated axons; however, the proteins required to organize and maintain this structure are not fully understood. Netrin-1 and its receptor Deleted in Colorectal Cancer (DCC) are proteins enriched at paranodes that are expressed by neurons and oligodendrocytes. To identify the specific function of DCC expressed by oligodendrocytes in vivo, we selectively eliminated DCC from mature myelinating oligodendrocytes using an inducible cre regulated by the proteolipid protein promoter. We demonstrate that DCC deletion results in progressive disruption of the organization of axonal domains, myelin ultrastructure, and myelin protein composition. Conditional DCC knock-out mice develop balance and coordination deficits and exhibit decreased conduction velocity. We conclude that DCC expression by oligodendrocytes is required for the maintenance and stability of myelin in vivo, which is essential for proper signal conduction in the CNS.
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Tamoxifen citrate for performance test, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tamoxifen, ≥99%
Supelco
Tamoxifen, analytical standard
Tamoxifen citrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Neurofilament M (145 kDa) Antibody, CT, serum, Chemicon®
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®