MilliporeSigma
  • Enhanced osteoblast proliferation and corrosion resistance of commercially pure titanium through surface nanostructuring by ultrasonic shot peening and stress relieving.
Enhanced osteoblast proliferation and corrosion resistance of commercially pure titanium through surface nanostructuring by ultrasonic shot peening and stress relieving.
The Journal of oral implantology (2014-07-16)
Shitu Jindal, Rajesh Bansal, Bijay P Singh, Rajiv Pandey, Shankar Narayanan, Mohan R Wani, Vakil Singh
ABSTRACT
This investigation was carried out to study the effect of a novel process of surface modification, surface nanostructuring by ultrasonic shot peening, on osteoblast proliferation and corrosion behavior of commercially pure titanium (c p-Ti) in simulated body fluid. A mechanically polished disc of c p-Ti was subjected to ultrasonic shot peening with stainless steel balls to create nanostructure at the surface. A nanostructure (<20 nm) with inhomogeneous distribution was revealed by atomic force and scanning electron microscopy. There was an increase of approximately 10% in cell proliferation, but there was drastic fall in corrosion resistance. Corrosion rate was increased by 327% in the shot peened condition. In order to examine the role of residual stresses associated with the shot peened surface on these aspects, a part of the shot peened specimen was annealed at 400°C for 1 hour. A marked influence of annealing treatment was observed on surface structure, cell proliferation, and corrosion resistance. Surface nanostructure was much more prominent, with increased number density and sharper grain boundaries; cell proliferation was enhanced to approximately 50% and corrosion rate was reduced by 86.2% and 41% as compared with that of the shot peened and the as received conditions, respectively. The highly significant improvement in cell proliferation, resulting from annealing of the shot peened specimen, was attributed to increased volume fraction of stabilized nanostructure, stress recovery, and crystallization of the oxide film. Increase in corrosion resistance from annealing of shot peened material was related to more effective passivation. Thus, the surface of c p-Ti, modified by this novel process, possessed a unique quality of enhancing cell proliferation as well as the corrosion resistance and could be highly effective in reducing treatment time of patients adopting dental and orthopedic implants of titanium and its alloys.
Product Description
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Titanium, rod, 200mm, diameter 2mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Titanium, microfoil, disks, 10mm, thinness 1.0μm, specific density 429μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, rod, 490mm, diameter 2mm, annealed, 99.6+%
Titanium, tube, 1220mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 25mm, diameter 16mm, as drawn, 99.99+%
Titanium, rod, 500mm, diameter 1.5mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 6mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 5mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%
Titanium, mesh, 100x100mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Titanium, rod, 100mm, diameter 2mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 9.5mm, annealed, 99.99+%
Titanium, rod, 100mm, diameter 2mm, as drawn, 99.99+%
Titanium, rod, 10mm, diameter 50mm, 99.99+%
Titanium, rod, 1000mm, diameter 3.0mm, annealed, 99.6+%