MilliporeSigma
  • Heme enzymes. Neutron cryo-crystallography captures the protonation state of ferryl heme in a peroxidase.
Heme enzymes. Neutron cryo-crystallography captures the protonation state of ferryl heme in a peroxidase.
Science (New York, N.Y.) (2014-07-12)
Cecilia M Casadei, Andrea Gumiero, Clive L Metcalfe, Emma J Murphy, Jaswir Basran, Maria Grazia Concilio, Susana C M Teixeira, Tobias E Schrader, Alistair J Fielding, Andreas Ostermann, Matthew P Blakeley, Emma L Raven, Peter C E Moody
ABSTRACT
Heme enzymes activate oxygen through formation of transient iron-oxo (ferryl) intermediates of the heme iron. A long-standing question has been the nature of the iron-oxygen bond and, in particular, the protonation state. We present neutron structures of the ferric derivative of cytochrome c peroxidase and its ferryl intermediate; these allow direct visualization of protonation states. We demonstrate that the ferryl heme is an Fe(IV)=O species and is not protonated. Comparison of the structures shows that the distal histidine becomes protonated on formation of the ferryl intermediate, which has implications for the understanding of O-O bond cleavage in heme enzymes. The structures highlight the advantages of neutron cryo-crystallography in probing reaction mechanisms and visualizing protonation states in enzyme intermediates.
Product Description
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
SAFC
L-Histidine
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Iron, foil, 6mm disks, thickness 0.004mm, 99.85%
Iron, foil, 50mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.004mm, 99.99+%
Iron, foil, 8mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.5mm, as rolled, 99.99+%
Iron, foil, light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Iron, foil, light tested, 50x50mm, thickness 0.025mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.005mm, 99.85%
Iron, foil, 50mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.020mm, 99.85%
Iron, foil, 5m coil, thickness 0.1mm, hard, 99.5%
Iron, foil, light tested, 50x50mm, thickness 0.038mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.007mm, 99.85%
Iron, foil, 6mm disks, thickness 0.01mm, 99.99+%
Iron, foil, 8mm disks, thickness 0.003mm, 99.85%
Iron, foil, 5m coil, thickness 0.075mm, hard, 99.5%