MilliporeSigma
Molar uprighting by a nickel-titanium spring based on a setup model.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics (2014-07-01)
MoonHee Kim, Minji Kim, Youn-Sic Chun
ABSTRACT
Molar uprighting is an important adjunctive treatment in orthodontics: repositioning the tilted molar eliminates the potentially pathologic condition and simplifies the ultimate restorative procedure. Although various methods for molar uprighting have been applied successfully, they still have some limitations and disadvantages in tooth movement. This article reports on a new clinical technique for molar uprighting with nickel-titanium springs based on a setup model. Two patients treated with this system are shown.
Product Description
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Nickel, foil, 2m coil, thickness 0.5mm, annealed, 99.98%
Nickel, foil, 150x300mm, thickness 0.25mm, annealed, 99.98%
Nickel, mesh, 30x30mm, nominal aperture 0.34mm, thickness 0.025mm, wire diameter 0.041mm, 70 wires/inch, open area 80%, electro-formed mesh, 99.9%
Titanium, rod, 200mm, diameter 2mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Titanium, microfoil, disks, 10mm, thinness 1.0μm, specific density 429μg/cm2, permanent mylar 3.5μm support, 99.6+%
Nickel, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Nickel, foil, not light tested, 300x300mm, thickness 0.0125mm, annealed, 99%
Titanium, rod, 490mm, diameter 2mm, annealed, 99.6+%
Nickel, wire reel, 10m, diameter 0.25mm, hard, 99.98%
Nickel, foil, 10mm disks, thickness 0.0075mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.038mm, 99.9%
Nickel, foil, 0.2m coil, thickness 0.004mm, hard, 99%
Titanium, tube, 1220mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%