MilliporeSigma
  • Seasonal induction of GABAergic excitation in the central mammalian clock.
Seasonal induction of GABAergic excitation in the central mammalian clock.
Proceedings of the National Academy of Sciences of the United States of America (2014-07-01)
Sahar Farajnia, Tirsa L E van Westering, Johanna H Meijer, Stephan Michel
ABSTRACT
The balance between excitation and inhibition is essential for the proper function of neuronal networks in the brain. The inhibitory neurotransmitter γ-aminobutyric acid (GABA) contributes to the network dynamics within the suprachiasmatic nucleus (SCN), which is involved in seasonal encoding. We investigated GABAergic activity and observed mainly inhibitory action in SCN neurons of mice exposed to a short-day photoperiod. Remarkably, the GABAergic activity in a long-day photoperiod shifts from inhibition toward excitation. The mechanistic basis for this appears to be a change in the equilibrium potential of GABA-evoked current. These results emphasize that environmental conditions can have substantial effects on the function of a key neurotransmitter in the central nervous system.
Product Description
Supelco
γ-Aminobutyric acid, analytical standard
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bumetanide, ≥98%
Bumetanide, European Pharmacopoeia (EP) Reference Standard