MilliporeSigma
[Whirlpool-dermatitis with "hot hands"].
Deutsche medizinische Wochenschrift (1946) (2014-07-02)
K-A Dietrich, T Ruzicka, T Herzinger
ABSTRACT
A 15-year-old boy presented with painful nodules on his palms and discrete pustules on the forearm. Two days earlier he had taken a bath in a new whirlpool. Bacteriological examination of the pustules revealed Pseudomonas aeruginosa. The cervical and axillary lymph nodes were inconspicuous. The dermal infection with Pseudomonas aeruginosa was treated with i.v. piperacillin 4 g/tazobactam 0,5 g twice daily. Furthermore, the patient received 400 mg ibuprofen twice daily per os. Seven days later all symptoms had resolved. Hot tubs are a potential source of cutaneous infections with the gram-negative rod Pseudomonas aeruginosa. The most common clinical manifestation of these infections is "hot tub- or jacuzzi-folliculitis" with pustules mainly on the trunk and lymphadenopathy. In children and adolescents, the clinical picture may differ, with a predominance of tender reddish nodules on the palms or soles named "hot hand/hot foot syndrome". In general, the first-line antibiotic in an infection with pseudomonas aeruginosa is ciprofloxacin but it is not recommended in children and adolescents.
Sigma-Aldrich
Piperacillin sodium salt, penicillin analog
Supelco
Piperacillin sodium salt, analytical standard