MilliporeSigma
  • Olaparib maintenance therapy in patients with platinum-sensitive relapsed serous ovarian cancer: a preplanned retrospective analysis of outcomes by BRCA status in a randomised phase 2 trial.
Olaparib maintenance therapy in patients with platinum-sensitive relapsed serous ovarian cancer: a preplanned retrospective analysis of outcomes by BRCA status in a randomised phase 2 trial.
The Lancet. Oncology (2014-06-03)
Jonathan Ledermann, Philipp Harter, Charlie Gourley, Michael Friedlander, Ignace Vergote, Gordon Rustin, Clare L Scott, Werner Meier, Ronnie Shapira-Frommer, Tamar Safra, Daniela Matei, Anitra Fielding, Stuart Spencer, Brian Dougherty, Maria Orr, Darren Hodgson, J Carl Barrett, Ursula Matulonis
ABSTRACT
Maintenance monotherapy with the PARP inhibitor olaparib significantly prolonged progression-free survival (PFS) versus placebo in patients with platinum-sensitive recurrent serous ovarian cancer. We aimed to explore the hypothesis that olaparib is most likely to benefit patients with a BRCA mutation. We present data from the second interim analysis of overall survival and a retrospective, preplanned analysis of data by BRCA mutation status from our randomised, double-blind, phase 2 study that assessed maintenance treatment with olaparib 400 mg twice daily (capsules) versus placebo in patients with platinum-sensitive recurrent serous ovarian cancer who had received two or more platinum-based regimens and who had a partial or complete response to their most recent platinum-based regimen. Randomisation was by an interactive voice response system, stratified by time to progression on penultimate platinum-based regimen, response to the most recent platinum-based regimen before randomisation, and ethnic descent. The primary endpoint was PFS, analysed for the overall population and by BRCA status. This study is registered with ClinicalTrials.gov, number NCT00753545. Between Aug 28, 2008, and Feb 9, 2010, 136 patients were assigned to olaparib and 129 to placebo. BRCA status was known for 131 (96%) patients in the olaparib group versus 123 (95%) in the placebo group, of whom 74 (56%) versus 62 (50%) had a deleterious or suspected deleterious germline or tumour BRCA mutation. Of patients with a BRCA mutation, median PFS was significantly longer in the olaparib group than in the placebo group (11·2 months [95% CI 8·3-not calculable] vs 4·3 months [3·0-5·4]; HR 0·18 [0·10-0·31]; p<0·0001); similar findings were noted for patients with wild-type BRCA, although the difference between groups was lower (7·4 months [5·5-10·3] vs 5·5 months [3·7-5·6]; HR 0·54 [0·34-0·85]; p=0·0075). At the second interim analysis of overall survival (58% maturity), overall survival did not significantly differ between the groups (HR 0·88 [95% CI 0·64-1·21]; p=0·44); similar findings were noted for patients with mutated BRCA (HR 0·73 [0·45-1·17]; p=0·19) and wild-type BRCA (HR 0·99 [0·63-1·55]; p=0·96). The most common grade 3 or worse adverse events in the olaparib group were fatigue (in ten [7%] patients in the olaparib group vs four [3%] in the placebo group) and anaemia (seven [5%] vs one [<1%]). Serious adverse events were reported in 25 (18%) patients who received olaparib and 11 (9%) who received placebo. Tolerability was similar in patients with mutated BRCA and the overall population. These results support the hypothesis that patients with platinum-sensitive recurrent serous ovarian cancer with a BRCA mutation have the greatest likelihood of benefiting from olaparib treatment. AstraZeneca.