MilliporeSigma
  • Estradiol modulates translocator protein (TSPO) and steroid acute regulatory protein (StAR) via protein kinase A (PKA) signaling in hypothalamic astrocytes.
Estradiol modulates translocator protein (TSPO) and steroid acute regulatory protein (StAR) via protein kinase A (PKA) signaling in hypothalamic astrocytes.
Endocrinology (2014-06-01)
Claire Chen, John Kuo, Angela Wong, Paul Micevych
ABSTRACT
The ability of the central nervous system to synthesize steroid hormones has wide-ranging implications for physiology and pathology. Among the proposed roles of neurosteroids is the regulation of the LH surge. This involvement in the estrogen-positive feedback demonstrates the integration of peripheral steroids with neurosteroids. Within the female hypothalamus, estradiol from developing follicles stimulates progesterone synthesis in astrocytes, which activate neural circuits regulating gonadotropin (GnRH) neurons. Estradiol acts at membrane estrogen receptor-α to activate cellular signaling that results in the release of inositol trisphosphate-sensitive calcium stores that are sufficient to induce neuroprogesterone synthesis. The purpose of the present studies was to characterize the estradiol-induced signaling leading to activation of steroid acute regulatory protein (StAR) and transporter protein (TSPO), which mediate the rate-limiting step in steroidogenesis, ie, the transport of cholesterol into the mitochondrion. Treatment of primary cultures of adult female rat hypothalamic astrocytes with estradiol induced a cascade of phosphorylation that resulted in the activation of a calcium-dependent adenylyl cyclase, AC1, elevation of cAMP, and activation of both StAR and TSPO. Blocking protein kinase A activation with H-89 abrogated the estradiol-induced neuroprogesterone synthesis. Thus, together with previous results, these experiments completed the characterization of how estradiol action at the membrane leads to the augmentation of neuroprogesterone synthesis through increasing cAMP, activation of protein kinase A, and the phosphorylation of TSPO and StAR in hypothalamic astrocytes.
Product Description
Supelco
Diethyl ether, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Diethyl ether, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
Progesterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%