MilliporeSigma
  • Should we consider topical silver nitrate irrigation as a definitive nonsurgical treatment for perianal fistula?
Should we consider topical silver nitrate irrigation as a definitive nonsurgical treatment for perianal fistula?
Diseases of the colon and rectum (2014-06-06)
Wafi Attaallah, Davut Tuney, Bahadir M Gulluoglu, M Umit Ugurlu, Omer Gunal, Cumhur Yegen
ABSTRACT
Surgery is currently the sole treatment modality for anal fistulas. However, surgery is associated with complications such as permanent incontinence, which reduces quality of life. To determine the rate of complete clinical healing of anal fistulas after irrigation of the fistula tract with silver nitrate solution as a nonsurgical treatment. Prospective single arm study. Tertiary university hospital : Adult patients with symptomatic perianal fistula presenting between June 2012 and January 2013. Irrigation of the fistula tract with 1% silver nitrate solution. Irrigation was repeated every 2 weeks when necessary. The primary outcome measure was the rate of complete clinical healing. Fifty-six consecutive patients with anal fistula were analyzed. Of those, 29 (52%) had complete clinical healing without recurrence for a median of 10 months. The median number of irrigations needed for complete clinical healing was 4 (1-10). The level of satisfaction was excellent in patients with complete clinical healing. The frequency of complaints was the only independent factor that had an impact on healing; patients with intermittent discharge had a significantly higher rate of complete clinical healing (67%) than those with continuous discharge (40%). There were no notable complications. Short follow-up, small sample size, and no comparisons. This study demonstrates that the application of silver nitrate solution often produces a favorable outcome in the treatment of anal fistula. This method may be considered as a first-line treatment for the disease because it is simple, performed on an outpatient basis, minimally invasive, and lacks the complications encountered with current conventional surgical modalities.
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O