MilliporeSigma
  • Segmental testicular infarction due to minocycline-induced antineutrophil cytoplasmic antibody--positive vasculitis.
Segmental testicular infarction due to minocycline-induced antineutrophil cytoplasmic antibody--positive vasculitis.
Urology (2014-05-06)
Timothy D Lyon, Matthew C Ferroni, Daniel P Casella, Louis A D'Agostino, Stephen V Jackman
ABSTRACT
Segmental testicular infarction is an uncommon clinical entity marked by acute scrotal pain and swelling. Classically, these appear as wedge-shaped, avascular, hypoechoic lesions on a testicular ultrasound. We present a unique case of testicular infarct caused by an antineutrophil cytoplasmic antibody-positive vasculitis secondary to the use of the antibiotic minocycline. The patient's symptoms resolved with cessation of minocycline. We suggest that patients who present with otherwise unexplained testicular infarction undergo a careful review of medications to uncover a potential cause.
Sigma-Aldrich
Minocycline hydrochloride, powder