MilliporeSigma
  • Resistance to irinotecan (CPT-11) activates epidermal growth factor receptor/nuclear factor kappa B and increases cellular metastasis and autophagy in LoVo colon cancer cells.
Resistance to irinotecan (CPT-11) activates epidermal growth factor receptor/nuclear factor kappa B and increases cellular metastasis and autophagy in LoVo colon cancer cells.
Cancer letters (2014-04-15)
Ming-Cheng Chen, Nien-Hung Lee, Tsung-Jung Ho, Hsi-Hsien Hsu, Chia-Hua Kuo, Wei-Wen Kuo, Yueh-Min Lin, Fuu-Jen Tsai, Chang-Hai Tsai, Chih-Yang Huang
ABSTRACT
Chemotherapy is usually applied to treat colon cancer but leads to chemoresistance, and increased metastasis and invasion. The main focus of this study is to observe effects of resistance to irinotecan (CPT-11) on metastasis, invasion and autophagy in CPT-11 resistant (CPT-11-R) LoVo colon cancer cells. CPT-11, a topoisomerase I inhibitor and a first-line chemotherapeutic drug, is used to treat colon cancer. CPT-11-R cells were constructed in a step-wise fashion with increasing CPT-11 doses. The CPT-11-R strain had a significantly lower expression of Wnt/β-catenin pathway, but induced an EGFR/IKKα/β/NF-κB pathway with elevated cell cycle, metastasis and basal autophagy.
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sigma-Aldrich
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder