MilliporeSigma
  • Effect of hydroxychloroquine on insulin sensitivity and lipid parameters in rheumatoid arthritis patients without diabetes mellitus: a randomized, blinded crossover trial.
Effect of hydroxychloroquine on insulin sensitivity and lipid parameters in rheumatoid arthritis patients without diabetes mellitus: a randomized, blinded crossover trial.
Arthritis care & research (2014-01-29)
Daniel H Solomon, Rajesh Garg, Bing Lu, Derrick J Todd, Emileigh Mercer, Tabatha Norton, Elena Massarotti
ABSTRACT
Observational studies suggest that hydroxychloroquine (HCQ) may reduce the risk of developing diabetes mellitus in patients with rheumatoid arthritis (RA). We examined the effect of HCQ on insulin resistance in subjects without diabetes mellitus with stable RA. Twenty-three RA subjects not currently using HCQ completed a 16-week, double-blind crossover study. Subjects were randomly allocated to receive HCQ (6.5 mg/kg/day) or placebo for the first 8 weeks, followed by crossover to the other arm for the final 8 weeks. Subjects underwent oral glucose tolerance testing and fasting lipid measurements at baseline, 8 weeks, and 16 weeks. The change ± SD from baseline in insulin sensitivity index (ISI), homeostatic model assessment for insulin resistance (HOMA-IR), and lipid parameters were compared between placebo and HCQ using linear regression. The mean patient age was 56 years, with 96% women, and the median body mass index was 26.0 kg/m2. After 8 weeks of HCQ, the mean ± SD ISI increase was 0.4 ± 2.9 compared with a small increase during placebo of 0.14 ± 3.1 (adjusted P = 0.785), and the mean ± SD HOMA-IR decrease was 0.3 ± 1.5 during HCQ versus a decrease of 0.42 ± 1.4 during placebo (adjusted P = 0.308). Small decreases in total cholesterol (12.7 mg/dl) and low-density lipoprotein (LDL) cholesterol (12.4 mg/dl) were observed during the HCQ treatment periods (both adjusted P < 0.05 compared to placebo). HCQ use for 8 weeks in patients without diabetes mellitus with stable RA produced no significant change in insulin resistance. We observed small and statistically significant improvements in total and LDL cholesterol during HCQ treatment.
Sigma-Aldrich
Hydroxychloroquine sulfate, ≥98% (HPLC), powder
Pricing and availability is not currently available.
USP
Hydroxychloroquine sulfate, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.