MilliporeSigma
  • Discovery of a series of 6,7-dimethoxy-4-pyrrolidylquinazoline PDE10A inhibitors.
Discovery of a series of 6,7-dimethoxy-4-pyrrolidylquinazoline PDE10A inhibitors.
Journal of medicinal chemistry (2007-01-19)
Thomas A Chappie, John M Humphrey, Martin P Allen, Kimberly G Estep, Carol B Fox, Lorraine A Lebel, Spiros Liras, Eric S Marr, Frank S Menniti, Jayvardhan Pandit, Christopher J Schmidt, Meihua Tu, Robert D Williams, Feng V Yang
ABSTRACT
A papaverine based pharmacophore model for PDE10A inhibition was generated via SBDD and used to design a library of 4-amino-6,7-dimethoxyquinazolines. From this library emerged an aryl ether pyrrolidyl 6,7-dimethoxyquinazoline series that became the focal point for additional modeling, X-ray, and synthetic efforts toward increasing PDE10A inhibitory potency and selectivity versus PDE3A/B. These efforts culminated in the discovery of 29, a potent and selective brain penetrable inhibitor of PDE10A.
Sigma-Aldrich
(S)-(+)-N-Boc-3-pyrrolidinol, 97%