MilliporeSigma
  • Structure-activity relationships of alpha, beta-unsaturated ketones as assessed by their cytotoxicity against oral tumor cells.
Structure-activity relationships of alpha, beta-unsaturated ketones as assessed by their cytotoxicity against oral tumor cells.
Anticancer research (2004-05-27)
Tohru Nakayachi, Eiji Yasumoto, Kensuke Nakano, Sufi Reza Md Morshed, Ken Hashimoto, Hirotaka Kikuchi, Hirofumi Nishikawa, Masami Kawase, Hiroshi Sakagami
ABSTRACT
A series of simple alpha, beta-unsaturated carbonyl compounds (1-26) was characterized for their cytotoxic profiles against oral human normal and tumor cells. Several cycloalkenones showed potent cytotoxic activities against human oral squamous cell carcinoma HSC-2 cell line. Among them, 4,4-dimethyl-2-cyclopenten-1-one (12) exhibited low cytotoxic activity against a normal human cell, gingival fibroblast HGF, and displayed higher tumor-specific cytotoxicity (SI value = CC50 (HGF)/CC50 (HSC-2) = 4.0). The cytotoxicities of the unsaturated lactones were moderately tumor-specific (SI = 1.5-1.9). Agarose gel electrophoresis showed that the induction of internucleosomal DNA fragmentation in human promyelocytic leukemia cell HL-60 is dependent on the structure of alpha, beta-unsaturated carbonyl compounds. Fluorometric protease assay showed that some, but not all compounds, activated the caspase 3 in a dose-dependent manner. All alpha, beta-unsaturated carbonyl compounds studied did not activate caspases 8 and 9. The cytotoxic activity of alpha, beta-unsaturated carbonyl compounds was profoundly reduced in the presence of N-acetylcysteine. The study suggests that the presence of a non sterically hindered Michael acceptor seems to be an essential structural requirement for the cytotoxic activity in alpha, beta-unsaturated ketones.
Sigma-Aldrich
Methyl 2-oxo-2H-pyran-3-carboxylate, 98%
Sigma-Aldrich
4,4-Dimethyl-2-cyclopenten-1-one, 99%