MilliporeSigma
  • Highly selective chemical modification of cruciform loops by diethyl pyrocarbonate.
Highly selective chemical modification of cruciform loops by diethyl pyrocarbonate.
Nucleic acids research (1986-05-27)
J C Furlong, D M Lilley
ABSTRACT
Diethyl pyrocarbonate reacts with the single-stranded loops of cruciform structures with great selectivity. Adenine bases are carbethoxylated, as a result of which the backbone may be cleaved with piperidine, and the level of chemical modification at each base may be determined. We have studied the ColE1 and (A-T)34 cruciforms of pColIR315 and pXG540. In each case we observe maximal modification at the most central adenosine of the loop, and an overall pattern of modification corresponding to a total loop size of about six bases. The results may be interpreted in terms of a model in which the loop has a defined tertiary structure. No modification was detected at either cruciform four-way junction, suggesting that this region is fully base-paired.
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)