MilliporeSigma
  • High-frequency genome editing using ssDNA oligonucleotides with zinc-finger nucleases.
High-frequency genome editing using ssDNA oligonucleotides with zinc-finger nucleases.
Nature methods (2011-07-19)
Fuqiang Chen, Shondra M Pruett-Miller, Yuping Huang, Monika Gjoka, Katarzyna Duda, Jack Taunton, Trevor N Collingwood, Morten Frodin, Gregory D Davis
ABSTRACT
Zinc-finger nucleases (ZFNs) have enabled highly efficient gene targeting in multiple cell types and organisms. Here we describe methods for using simple ssDNA oligonucleotides in tandem with ZFNs to efficiently produce human cell lines with three distinct genetic outcomes: (i) targeted point mutation, (ii) targeted genomic deletion of up to 100 kb and (iii) targeted insertion of small genetic elements concomitant with large genomic deletions.
Product Description
Custom DNA Oligos, Design and Order Custom Oligos
Pricing and availability is not currently available.