MilliporeSigma
Synthesis and anti-inflammatory activity of resveratrol analogs.
Chemical & pharmaceutical bulletin (2005-12-06)
Guoliang Chen, Wei Shan, Yingliang Wu, Lixiang Ren, Jinhua Dong, Zhizhong Ji
ABSTRACT
Seventeen novel resveratrol derivatives were synthesized. Their anti-inflammatory activities were tested on xylene-induced mouse ear edema. The pharmacological results showed that some compounds have potent anti-inflammatory activities.