MilliporeSigma
  • Synthesis, characterization, and molecular docking analysis of novel benzimidazole derivatives as cholinesterase inhibitors.
Synthesis, characterization, and molecular docking analysis of novel benzimidazole derivatives as cholinesterase inhibitors.
Bioorganic chemistry (2013-07-28)
Yeong Keng Yoon, Mohamed Ashraf Ali, Ang Chee Wei, Tan Soo Choon, Kooi-Yeong Khaw, Vikneswaran Murugaiyah, Hasnah Osman, Vijay H Masand
ABSTRACT
Two series of novel acetylcholinesterase (AChE) and butyrylcholinesterase (BChE) inhibitors containing benzimidazole core structure were synthesized by a four-step reaction pathway starting from 4-fluoro-3-nitrobenzoic acid as the basic compound. The structure of the novel benzimidazoles was characterized and confirmed by the elemental and mass spectral analyses as well as (1)H NMR spectroscopic data. Of the 34 novel synthesized compounds, three benzimidazoles revealed AChE inhibition with IC50<10 μM. The highest inhibitory activity (IC50=5.12 μM for AChE and IC50=8.63 μM for BChE) corresponds to the compound 5IIc (ethyl 1-(3-(1H-imidazol-1-yl)propyl)-2-(4-nitrophenyl)-1H-benzo[d]imidazole-5-carboxylate). The relationship between lipophilicity and the chemical structures as well as their limited structure-activity relationship was discussed.
Sigma-Aldrich
4-Fluoro-3-nitrobenzoic acid, 98%