MilliporeSigma
  • Synthesis and microbiological activity of some novel 5-benzamido- and 5-phenylacetamido-substituted 2-phenylbenzoxazole derivatives.
Synthesis and microbiological activity of some novel 5-benzamido- and 5-phenylacetamido-substituted 2-phenylbenzoxazole derivatives.
Farmaco (Societa chimica italiana : 1989) (2000-09-13)
E A Sener, O T Arpaci, I Yalçin, N Altanlar
ABSTRACT
The synthesis and microbiological activity of a new series of 5-benzamido- and 5-phenylacetamidosubstituted-2-phenylbenzoxazole derivatives (1-26) were described. The in vitro microbiological activity of the compounds was determined against Gram-positive, Gram-negative bacteria and the yeast Candida albicans in comparison with standard drugs. Microbiological results indicated that the synthesized compounds possessed a broad spectrum of activity against the tested microorganisms. The compounds 1, 21, 25 showed higher activity than tetracycline and streptomycin against Pseudomonas aeruginosa.