MilliporeSigma
  • Chirally deuterated benzyl chlorides from benzyl alcohols via hexachloroacetone/polymer-supported triphenylphosphine: synthesis of protected (2S, 3S)-[3-(2)H, (15)N]-tyrosine.
Chirally deuterated benzyl chlorides from benzyl alcohols via hexachloroacetone/polymer-supported triphenylphosphine: synthesis of protected (2S, 3S)-[3-(2)H, (15)N]-tyrosine.
Journal of labelled compounds & radiopharmaceuticals (2013-11-29)
Derek W Barnett, Maryanne S Refaei, Robert W Curley
ABSTRACT
Chirally deuterated benzyl chlorides were prepared using novel, general hexachloroacetone/polymer-supported triphenylphosphine treatment of chirally deuterated benzyl alcohols. Doubly labeled protected tyrosine was obtained in 62% yield with 86% de at the α-carbon and 82% de at the β-carbon. Key in the synthesis was the alkylation of (15)N-labeled (-)-8-phenylmenthylhippurate with R-(-)-4-triisopropylsilyloxybenzyl-α-d chloride.
Sigma-Aldrich
Hexachloro-2-propanone, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Vinyl laurate, Wacker Chemie AG, ≥98% (GC)