MilliporeSigma
  • A new potential attractant for Anastrepha obliqua from Spondias mombin fruits.
A new potential attractant for Anastrepha obliqua from Spondias mombin fruits.
Journal of chemical ecology (2006-03-24)
Leopoldo Cruz-López, Edi A Malo, Jorge Toledo, Armando Virgen, Alejandro Del Mazo, Julio C Rojas
ABSTRACT
Nonirradiated males and females of Anastrepha obliqua (Macquart) were attracted to and landed more frequently on ripe fruits of Spondias mombin L. than on artificial fruit in wind tunnel bioassays. Porapak Q volatile extracts of S. mombin were also attractive and elicited landing on artificial fruit for both sexes. Combined gas chromatographic-electroantennographic detection (GC-EAD) analysis of volatile extracts showed that nine volatile compounds elicited repeatable antennal responses from females and males. The EAD-active compounds were identified by GC-mass spectrometry (MS) as follows: ethyl butyrate, isopropyl butyrate, hexan-1-ol, propyl butyrate, isobutyl butyrate, ethyl hexanoate, isopentyl butyrate, ethyl benzoate, and ethyl octanoate. In wind tunnel bioassays, males and females were attracted and landed more frequently on lures containing the nine-component blend of synthetic compounds than on unscented controls. Field cage bioassays showed that multilure traps baited with the nine-synthetic blend captured significantly more A. obliqua than traps baited with hydrolyzed protein or water.
Sigma-Aldrich
Isopropyl butyrate, 99%