MilliporeSigma
  • Mononucleotide recognition by cyclic trinuclear palladium(II) complexes containing 4,7-phenanthroline N,N bridges.
Mononucleotide recognition by cyclic trinuclear palladium(II) complexes containing 4,7-phenanthroline N,N bridges.
Dalton transactions (Cambridge, England : 2003) (2004-07-15)
Miguel A Galindo, Jorge A R Navarro, M Angustias Romero, Miguel Quirós
ABSTRACT
Reaction of [(dach)Pd(NO3)2] entities (dach = (R,R)-1,2-diaminocyclohexane, (S,S)-1,2-diaminocyclohexane) and 4,7-phenanthroline (phen) providing, respectively, 90 and 120 degrees bond angles, leads to the formation of two novel positively charged homochiral cyclic trinuclear metallacalix[3]arene species [((R,R)-1,2-diaminocyclohexane)Pd(phen)]3(NO3)6 (2a) and [((S,S)-1,2-diaminocyclohexane)Pd(phen)]3(NO3)6 (2b). These species have been characterised by 1)H NMR and X-ray diffraction methods (2b), showing that they possess accessible cavities suited for supramolecular recognition processes. We prove, indeed, from 1H NMR studies the inclusion of mononucleotides inside the cavity of the trinuclear species [(ethylenediamino)Pd(phen)]3(6+) (1), [((R,R)-1,2-diaminocyclohexane)Pd(phen)]3(6+) (2a) and [((S,S)-1,2-diaminocyclohexane)Pd(phen)]3(6+) (2b) in aqueous solution. Association constants (K(ass)) range from 85 +/- 6 M(-1) for the interaction between [(ethylenediamine)Pd(phen)]3(6+) and adenosine monophosphate to 37 +/- 4 M(-1) for the interaction between [(1,2-diaminocyclohexane)Pd(phen)]3(6+) and thymidine monophosphate. We invoke the synergy of electrostatic, anion-pi and pi-pi interactions to explain the recognition of mononucleotides inside the cavity of the metallacalix[3]arenes.
Sigma-Aldrich
4,7-Phenanthroline, 98%