MilliporeSigma
Solid ionic matrixes for direct tissue analysis and MALDI imaging.
Analytical chemistry (2006-02-02)
R Lemaire, J C Tabet, P Ducoroy, J B Hendra, M Salzet, I Fournier
ABSTRACT
Direct analysis of tissue by MALDI-MS allows the acquisition of its biomolecular profile while maintaining the integrity of the tissue, giving cellular localization, and avoiding tedious extraction and purification steps. However, direct tissue analysis generally leads to some extent to a lowered spectral quality due to variation in thickness, freezing tissue date, and nature of the tissue. We present here new technical developments for the direct tissue analysis of peptides with ionic liquid made of matrix mixtures (alpha-cyano-4-hydroxycinnamic acid (CHCA)/2-amino-4-methyl-5-nitropyridine and alpha-cyano-4-hydroxycinnamic acid/N,N-dimethylaniline (CHCA/DANI)). The properties of these direct tissue analysis matrixes, especially CHCA/aniline when compared to CHCA, 2,5-dihydroxybenzoic acid, and sinapinic acid, are as follows: (1) better spectral quality in terms of resolution, sensitivity, intensity, noise, number of compounds detected, and contaminant tolerance, (2) better crystallization on tissues, i.e., coverage capacity, homogeneity of crystallization, homogeneity of crystal sizes, and time of crystallization, (3) better analysis duration in term of vacuum stability, (4) better resistance to laser irradiation especially for high-frequency lasers, (5) better ionic yield in negative mode, and (6) enough fragmentation yield to use the PSD mode on sections to get structural information. Applied to MALDI imaging on a MALDI LIFT-TOF with a 50-Hz laser frequency, these ionic matrixes have allowed the realization of a new type of image in both polarities and reflector mode using the same tissue section. These results give a new outlook on peptide tissue profiling by MS, characterization of compounds from tissue slices, and MALDI-MS high-quality imaging.
Sigma-Aldrich
2-Amino-4-methyl-5-nitropyridine, 98%