MilliporeSigma
Antifungal activity of substituted nitrobenzenes and anilines.
Applied microbiology (1971-09-01)
H Gershon, M W McNeil, R Parmegiani, P K Godfrey
ABSTRACT
Series of 1,3-dihalogeno-5-nitrobenzenes, 3- and 3,5-halogenoanilines, and 2,6-dihalogeno-4-nitroanilines were tested for fungitoxicity against Aspergillus niger, A. oryzae, Trichoderma viride, Myrothecium verrucaria, and Trichophyton mentagrophytes in shaken culture by using Sabouraud dextrose broth enriched with yeast extract as the test medium. 1,3-Dichloro-5-nitrobenzene, 1,3-dibromo-5-nitrobenzene, 3-iodoaniline, 3,5-dichloroaniline, and 3,5-dibromoaniline were found to possess sufficient activity, compared with 8-quinolinol, to warrant further study.