MilliporeSigma
Preclinical pharmacology of metrifonate.
Pharmacotherapy (1998-04-16)
M W Jann
ABSTRACT
Alzheimer's disease is a chronic neurodegenerative disorder that is characterized by memory impairment, cognitive dysfunction, behavioral disturbances, and deficits in activities of daily living. A consistent observation in these patients is that cholinergic neurons are affected and deteriorate over time, leading to decreased levels of acetylcholine (ACh). Acetylcholinesterase (AChE) inhibitors, which attempt to prevent the breakdown of ACh, may be classified as short acting, intermediate acting, and long acting based on AChE regeneration time. Metrifonate is converted by a nonenzymatic process to the long-acting cholinesterase inhibitor 2,2-dichlorovinyl dimethyl phosphate (DDVP). Acetylcholinesterase inhibition produced by metrifonate occurs rapidly, is dose dependent, can be detected by inhibition measured in red blood cells, and can be reversed by oxime administration. Metrifonate and DDVP improved performance in young rats; cognitive improvement in aged rats also was observed. Both agents were well tolerated and did not have significant effects on various preclinical pharmacologic safety tests.
Product Description
Supelco
Trichlorfon, PESTANAL®, analytical standard