MilliporeSigma
Biological effects of myricetin.
General pharmacology (1997-08-01)
K C Ong, H E Khoo
ABSTRACT
1. Myricetin is a natural bioflavonoid whose occurrence in nature is widespread among plants. 2. It has been demonstrated to possess both antioxidative properties and prooxidative properties. 3. It is a potent anticarcinogen and antimutagen, although it has been shown to promote mutagenesis with the use of the Ames Test. 4. Its therapeutic potential and benefits in cardiovascular diseases and diabetes mellitus also are reviewed.
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Supelco
Myricetin, analytical standard