MilliporeSigma
  • Modulation of human polymorphonuclear neutrophil function by macrolides: preliminary data concerning dirithromycin.
Modulation of human polymorphonuclear neutrophil function by macrolides: preliminary data concerning dirithromycin.
The Journal of antimicrobial chemotherapy (1993-03-01)
M T Labro, J el Benna, H Abdelghaffar
ABSTRACT
Polymorphonuclear neutrophils (PMN) play a prominent role in the host response to infectious diseases. One major bactericidal mechanism used by these cells is the production of reactive oxygen species during what is referred to as the oxidative burst. However, excessive oxidant generation can also be involved in cell and tissue damage associated with severe inflammatory reactions. Macrolide antibiotics are able to penetrate and concentrate within phagocytes and have been successfully used to treat infections due to facultative intracellular pathogens. However, intracellular accumulation of macrolides with possible alkalinization of cellular compartments may interfere with normal cell function. In-vitro and ex-vivo data suggest that macrolides affect various phagocytic functions. This paper presents an overview of the published data concerning the modulation of neutrophil function by macrolides. Preliminary data concerning the in-vitro modulation of the neutrophil oxidative burst by dirithromycin and its metabolite, erythromycylamine, are also discussed.
Product Description
Dirithromycin, European Pharmacopoeia (EP) Reference Standard