MilliporeSigma
Pathogenetic studies of hexane and carbon disulfide neurotoxicity.
Critical reviews in toxicology (1995-01-01)
D G Graham, V Amarnath, W M Valentine, S J Pyle, D C Anthony
ABSTRACT
Two commonly employed solvents, n-hexane and carbon disulfide (CS2), although chemically dissimilar, result in identical neurofilament-filled swellings of the distal axon in both the central and peripheral nervous systems. Whereas CS2 is itself a neurotoxicant, hexane requires metabolism to the gamma-diketone, 2,5-hexanedione (HD). Both HD and CS2 react with protein amino functions to yield initial adducts (pyrrolyl or dithiocarbamate derivatives, respectively), which then undergo oxidation or decomposition to an electrophile (oxidized pyrrole ring or isothiocyanate), that then reacts with protein nucleophiles to result in protein cross-linking. It is postulated that progressive cross-linking of the stable neurofilament during its anterograde transport in the longest axons ultimately results in the accumulation of neurofilaments within axonal swellings. Reaction with additional targets appears to be responsible for the degeneration of the axon distal to the swellings.
Sigma-Aldrich
Carbon disulfide, anhydrous, ≥99%
Sigma-Aldrich
Carbon disulfide, suitable for IR spectroscopy, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (GC)
Sigma-Aldrich
Carbon disulfide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Carbon disulfide, ReagentPlus®, purified by redistillation, ≥99.9%
Sigma-Aldrich
Carbon disulfide, ACS reagent, ≥99.9%
Sigma-Aldrich
Carbon disulfide, ReagentPlus®, low benzene, ≥99.9%
Sigma-Aldrich
Carbon disulfide, ACS reagent, ≥99.9%
Supelco
Carbon disulfide solution, certified reference material, 5000 μg/mL in methanol