MilliporeSigma
  • Cobalamin-dependent methionine synthase: the structure of a methylcobalamin-binding fragment and implications for other B12-dependent enzymes.
Cobalamin-dependent methionine synthase: the structure of a methylcobalamin-binding fragment and implications for other B12-dependent enzymes.
Current opinion in structural biology (1994-12-01)
C L Drennan, R G Matthews, M L Ludwig
ABSTRACT
Cobalamin-dependent methionine synthase is a large enzyme composed of structurally and functionally distinct regions. Recent studies have begun to define the roles of several regions of the protein. In particular, the structure of a 27 kDa cobalamin-binding fragment of the enzyme from Escherichia coli has been determined by X-ray crystallography, and has revealed the motifs and interactions responsible for recognition of the cofactor. The amino acid sequences of several adenosylcobalamin-dependent enzymes, the methylmalonyl coenzyme A mutases and glutamate mutases, show homology with the cobalamin-binding region of methionine synthase and retain conserved residues that are determinants for the binding of the prosthetic group, suggesting that these mutases and methionine synthase share common three-dimensional structures.
Sigma-Aldrich
Methylcobalamin, vitamin B12 analog