MilliporeSigma
Two cases of lethal nitrazepam poisoning.
The American journal of forensic medicine and pathology (1995-06-01)
I Brødsgaard, A C Hansen, A Vesterby
ABSTRACT
This case report describes two cases of lethal poisoning caused by a combination of advanced chronic disease and an overdose of nitrazepam. In both cases, a relatively small blood concentration of nitrazepam was found postmortem.
Product Description
Supelco
Nitrazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®