MilliporeSigma
  • Severe hypoalphalipoproteinemia induced by a combination of probucol and clofibrate.
Severe hypoalphalipoproteinemia induced by a combination of probucol and clofibrate.
Advances in experimental medicine and biology (1986-01-01)
J Davignon, A C Nestruck, P Alaupovic, D Bouthillier
ABSTRACT
Observation of a markedly depressed HDL-cholesterol (5 mg/dL) in a patient with familial hypercholesterolemia (FH) receiving probucol (1 g/day) and clofibrate (2 g/day) prompted a review of all cases treated by this combination at our lipid clinic. Hypoalphalipoproteinemia (HDL-C less than 15 mg/dL) developed in 19 of 28 (70%) hyperlipidemic subjects who received this combination for an average of 1.5 years. This effect was sustained and reversible; it did not occur on either drug alone and was manifested on average 17 weeks after the combination was started. Plasma triglycerides increased significantly in most of those patients susceptible to this reduction in HDL-C. Plasma apolipoprotein A-I was decreased 82%, in proportion to the HDL-C fall, whereas apo A-II was lowered 65%. Since apo C-III concentrations tended to be high, the apo A-I/C-III ratio was markedly depressed. Apo E levels were unchanged and apo B levels reflected the high LDL concentrations of the underlying disease. An intermediate response was observed in subjects whose HDL-C remained well above 15 mg/dL on the combination. No deleterious side-effects could be attributed directly to the administration of the combined drugs in this high-risk group. One patient actually showed complete regression of xanthelasma and extensor tendon xanthomas of the finger on the combination. A parallel is drawn with Fish-Eye disease and the presence of the apolipoprotein A-I Milano variant, where similar HDL-C levels are observed in the absence of an increased atherogenic risk. It is mandatory to monitor plasma HDL-C in hypercholesterolemic patients treated with this combination, otherwise the pronounced HDL-deficiency could go unnoticed.
Product Description
Clofibrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Clofibrate, analytical standard
Sigma-Aldrich
Clofibrate, liquid