MilliporeSigma
  • Balancing risks and benefits of therapy for patients with favorable-risk limited-stage Hodgkin lymphoma: the role of doxorubicin, bleomycin, vinblastine, and dacarbazine chemotherapy alone.
Balancing risks and benefits of therapy for patients with favorable-risk limited-stage Hodgkin lymphoma: the role of doxorubicin, bleomycin, vinblastine, and dacarbazine chemotherapy alone.
Hematology/oncology clinics of North America (2013-11-30)
Annette E Hay, Ralph M Meyer
ABSTRACT
Because long-term survival of patients with nonbulky stage IA to IIA Hodgkin lymphoma is dependent on disease control and avoidance of late toxic effects associated with the treatment received, the initial choice of treatment can be associated with trade-offs that balance optimum disease control with avoidance of these late effect risks. Health professionals and patients face the dilemma of making treatment decisions without the benefit of completely understanding the risk-benefit balances associated with how current treatments affect all outcomes of interest. Optimum management of these patients requires careful multidisciplinary evaluation and communication strategies that account for patient preferences.
Sigma-Aldrich
Dacarbazine, antineoplastic purine analog
Pricing and availability is not currently available.
Dacarbazine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.