MilliporeSigma
Fluorescent silver nanoclusters as DNA probes.
Nanoscale (2013-07-06)
Judy M Obliosca, Cong Liu, Hsin-Chih Yeh
ABSTRACT
Fluorescent silver nanoclusters (few atoms, quantum sized) have attracted much attention as promising substitutes for conventional fluorophores. Due to their unique environmental sensitivities, new fluorescent probes have been developed based on silver nanoclusters for the sensitive and specific detection of DNA. In this review we present the recent discoveries of activatable and color-switchable properties of DNA-templated silver nanoclusters and discuss the strategies to use these new properties in DNA sensing.
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Silver, foil, light tested, 50x50mm, thickness 0.02mm, 99.97%
Silver, foil, light tested, 50x50mm, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95+%
Silver, foil, 10m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, light tested, 100x100mm, thickness 0.02mm, 99.97%
Silver, foil, 5m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, not light tested, 100x100mm, thickness 0.006mm, 99.97%
Silver, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 1m coil, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 2m coil, thickness 0.0125mm, 99.95+%
Silver, wire reel, 500m, diameter 0.05mm, annealed, 99.99%
Silver, wire reel, 1m, diameter 0.015mm, hard, 99.99%
Silver, wire reel, 10m, diameter 0.05mm, annealed, 99.99%