MilliporeSigma
  • Phentermine and topiramate extended-release: a new treatment for obesity and its role in a complications-centric approach to obesity medical management.
Phentermine and topiramate extended-release: a new treatment for obesity and its role in a complications-centric approach to obesity medical management.
Expert opinion on drug safety (2013-06-07)
William Timothy Garvey
ABSTRACT
Weight-management options include lifestyle modifications, bariatric surgery and, until recently, limited pharmacotherapy. Phentermine and topiramate extended-release (phentermine/topiramate ER) has recently been approved in the USA for chronic weight management in obese adults and overweight adults with weight-related co-morbidities in conjunction with a reduced-calorie diet and increased physical activity. This review describes the pharmacology and clinical trials data for phentermine/topiramate ER and its role in a complications-centric approach to medical care of the overweight and obese patient. Phentermine/topiramate ER is an effective and safe weight-loss medication that can produce and sustain approximately 10% loss of body weight. This is a landmark development in the pharmacotherapy of obesity. By offering an effective medical option to complement lifestyle and surgical approaches, phentermine/topiramate ER enables a comprehensive medical model for obesity care. The overall approach to the overweight and obese patient should be to identify individuals who will benefit most from therapy based on cardiometabolic or mechanical complications, establish therapeutic targets and goals for ameliorating these complications and selecting the treatment modality and intensity for weight loss to achieve these goals. This complications-centric model emphasizes weight loss as a tool to ameliorate obesity-related complications and optimizes benefit/risk for achieving the best outcomes in overweight/obese patients.
Product Description
Supelco
Topiramate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Topiramate, ≥98% (HPLC), solid
USP
Topiramate, United States Pharmacopeia (USP) Reference Standard