MilliporeSigma
  • Fenofibrate: a new treatment for diabetic retinopathy. Molecular mechanisms and future perspectives.
Fenofibrate: a new treatment for diabetic retinopathy. Molecular mechanisms and future perspectives.
Current medicinal chemistry (2013-06-12)
Rafael Simó, Sayon Roy, Francine Behar-Cohen, Anthony Keech, Paul Mitchell, Tien Yin Wong
ABSTRACT
Despite improving standards of care, people with diabetes remain at risk of development and progression of diabetic retinopathy (DR) and visual impairment. Identifying novel therapeutic approaches, preferably targeting more than one pathogenic pathway in DR, and at an earlier stage of disease, is attractive. There is now consistent evidence from two major trials, the Fenofibrate Intervention and Event Lowering in Diabetes (FIELD) study and the Action to Control Cardiovascular Risk in Diabetes Eye (ACCORD-Eye) study, totalling 11,388 people with type 2 diabetes (5,701 treated with fenofibrate) that fenofibrate reduces the risk of development and progression of DR. Therefore, fenofibrate may be considered a preventive strategy for patients without DR or early intervention strategy for those with mild DR. A number of putative therapeutic mechanisms for fenofibrate, both dependent and independent of lipids, have been proposed. A deeper understanding of the mode of action of fenofibrate will further help to define how best to use fenofibrate clinically as an adjunct to current management of DR.
Sigma-Aldrich
Fenofibrate, ≥99%, powder
USP
Fenofibrate, United States Pharmacopeia (USP) Reference Standard
Fenofibrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Fenofibrate, Pharmaceutical Secondary Standard; Certified Reference Material