MilliporeSigma
Toluene abuse and white matter: a model of toxic leukoencephalopathy.
The Psychiatric clinics of North America (2013-05-22)
Christopher M Filley
ABSTRACT
The brain is the primary target of toluene (methylbenzene), the major solvent in spray paint and a constituent of many other easily obtained commercial and industrial products. The effects of acute intoxication can be dramatic and the lasting adverse effects of inhalants may be highly injurious. Mental status alterations range from acute confusional state to coma. Toluene abuse effects on white matter are demonstrable neuroradiologically and neuropathologically, and have important neurobehavioral consequences.
Product Description
Supelco
Toluene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)