MilliporeSigma
  • Inactivation of deoxynivalenol-contaminated cereal grains with sodium metabisulfite: a review of procedures and toxicological aspects.
Inactivation of deoxynivalenol-contaminated cereal grains with sodium metabisulfite: a review of procedures and toxicological aspects.
Mycotoxin research (2013-04-23)
Sven Dänicke, Susanne Kersten, Hana Valenta, Gerhard Breves
ABSTRACT
Both hydrothermal treatment and wet preservation of mainly deoxynivalenol (DON)-containing, Fusarium toxin (FUS)-contaminated cereal grains with sodium metabisulfite (Na2S2O5 [SBS]) were successfully demonstrated to reduce the DON contamination through formation of the sulfonated derivative of DON, termed as DON sulfonate (DONS). The wet preservation is particularly interesting from a practical viewpoint as it can be easily performed at the farm level where the cereal grains are harvested and utilized in pig feeding. This review compiles the literature with regard to the chemical characterization and the detection of DONS, technical procedures and their efficacies, toxicological aspects and toxic effects of DON, DONS and SBS, and detection of DONS, DON and further metabolites in physiological specimens of pigs.
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
Supelco
Sodium Metabisulfite, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium metabisulfite, puriss. p.a., ACS reagent, reag. Ph. Eur., dry, 98-100.5%
Sigma-Aldrich
Sodium metabisulfite, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium metabisulfite, reagent grade, 97%
Sigma-Aldrich
Sodium metabisulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Sigma-Aldrich
Sodium metabisulfite, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Deoxynivalenol
Supelco
Deoxynivalenol solution, 100 μg/mL in acetonitrile, analytical standard
4-Deoxynivalenol in acetonitrile, IRMM®, certified reference material