MilliporeSigma
  • Haemostasis in neurosurgery: what is the evidence for gelatin-thrombin matrix sealant?
Haemostasis in neurosurgery: what is the evidence for gelatin-thrombin matrix sealant?
Journal of clinical neuroscience : official journal of the Neurosurgical Society of Australasia (2013-02-07)
Henry H I Yao, Matthew K H Hong, Katharine J Drummond
ABSTRACT
Strict intra-operative haemostasis is essential in the practice of neurosurgery. Over the last century, haemostatic methods have advanced significantly and the modern surgeon is now faced with an array of haemostatic agents, each with subtly different qualities and proven in different contexts with various levels of evidence. The popularity of endoscopic and laparoscopic procedures in other surgical specialties has encouraged the introduction of novel agents to achieve haemostasis where conventional methods have proven difficult. These agents are beginning to find a role in routine use for surgery in both the elective and emergent settings. This article reviews the mechanisms of different haemostasis methods and the current evidence for their use in neurosurgery, with a focus on the more recently introduced gelatin-thrombin matrix sealant (Floseal [Baxter, Hayward, CA, USA]).
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture