MilliporeSigma
  • Palinopsia induced by topiramate and zonisamide in a patient with migraine.
Palinopsia induced by topiramate and zonisamide in a patient with migraine.
Clinical neuropharmacology (2013-03-19)
Fernando Sierra-Hidalgo, Eduardo de Pablo-Fernández
ABSTRACT
Palinopsia is an illusory visual phenomenon consisting in the persistence or recurrence of visual images after the exciting stimulus object has been removed. It has been reported in association with parietal and occipital lobe lesions, migraine auras, and related to the use of several drugs and illicit drugs. Here, we report the case of a 23-year-old woman with a 4-year history of episodic migraine with aura who developed palinopsia during sequential prophylactic therapies with topiramate and zonisamide. Although the exact physiopathology of this phenomenon remains unknown, topiramate- and zonisamide-induced palinopsia support an increase on serotonergic activity as a possible mechanism for this visual disorder.
Product Description
Supelco
Topiramate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Topiramate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Topiramate, ≥98% (HPLC), solid